Invitrogen
Manufacturer Code:PA5115145
Catalog # PIPA5115145
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human DSP corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2300002E22Rik 250/210 kDa paraneoplastic pemphigus antigen 5730453H04Rik AA407887 AA407888 AW109828 DCWHKTA Desmoplakin desmoplakin I/II Desmoplakin II Desmoplakin-I desmosomal cytoskeletal connector molecule DP Dsp rul; 250/210 kDa paraneoplastic pemphigus antigen; Desmoplakin; desmoplakin I/II; desmosomal cytoskeletal connector molecule; DP
Gene Aliases: 2300002E22Rik; 5730453H04Rik; AA407887; AA407888; AW109828; DCWHKTA; DP; DSP; rul
UniProt ID: (Human) P15924, (Mouse) E9Q557
Entrez Gene ID: (Human) 1832, (Mouse) 109620
Molecular Function:
actin family cytoskeletal protein
cytoskeletal protein
non-motor actin binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.