Invitrogen
Manufacturer Code:PA563728
Catalog # PIPA563728
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human Desmoglein 4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cadherin family member 13; Desmoglein-4; Desmoglein-4 Desmoglein4 Cadherin family member 13 DSG4 CDHF13
Gene Aliases: CDGF13; CDHF13; DSG4; HYPT6; LAH
UniProt ID: (Human) Q86SJ6
Entrez Gene ID: (Human) 147409
Molecular Function:
cadherin
cell adhesion molecule
cell junction protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.