Novus Biologicals
Manufacturer Code:NB1001643AF405
Catalog # NB1001643W
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 5G11 |
Immunogen | Human Desmoglein 3 recombinant protein [UniProt# P32926] |
Conjugate | Alexa Fluor 405 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 130 kDa pemphigus vulgaris antigen; Cadherin family member 6; Cadherin family member 6 CDHF6130-kD pemphigus vulgaris antigen desmoglein 3 desmoglein 3 (pemphigus vulgaris antigen) desmoglein-3 DKFZp686P23184130 kDa pemphigus vulgaris antigen pemphigus vulgaris antigen PVA; Desmoglein-3; PVA
Gene Aliases: CDHF6; DSG3; PVA
UniProt ID: (Human) P32926
Entrez Gene ID: (Human) 1830
Molecular Function: cadherin cell adhesion molecule cell junction protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.