Invitrogen
Manufacturer Code:MA530494
Catalog # PIMA530494
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 4 |
Immunogen | Recombit Human DSC2 Protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: ARVD11 CDHF2 DG2 DGII/III DSC3 cadherin family member 2 desmocollin-2 desmosomal glycoprotein II desmosomal glycoprotein II/III desmosomal glycoprotein III; Cadherin family member 2; Desmocollin-2; Desmocollin-3; Desmosomal glycoprotein II; desmosomal glycoprotein II/III; Desmosomal glycoprotein III
Gene Aliases: ARVD11; CDHF2; DG2; DGII/III; DSC2; DSC3
UniProt ID: (Human) Q02487
Entrez Gene ID: (Human) 1824
Molecular Function: cadherin cell adhesion molecule cell junction protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.