Invitrogen
Manufacturer Code:14974782
Catalog # 50-112-2816
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | DE-U-10 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CMD1I CSM2 DES FLJ12025; Desmin; desmin-like protein; intermediate filament protein; muscle-specific intermediate filament desmin; muscle-specific intermediate filament protein; mutant desmin p.K241E
Gene Aliases: CSM1; CSM2; DES; I79_018932; LGMD2R
UniProt ID: (Bovine) O62654, (Pig) P02540, (Human) P17661, (Rat) P48675, (Mouse) P31001
Entrez Gene ID: (Sheep) 101117308, (Bovine) 280765, (Pig) 396725, (Rabbit) 100328890, (Chicken) 395906, (Goat) 102180712, (Human) 1674, (Rat) 64362, (Mouse) 13346, (Chinese hamster) 100764990
Molecular Function: cytoskeletal protein intermediate filament structural protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.