Novus Biologicals
Manufacturer Code:NB120171560.5ML
Catalog # NB120171565
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | D33 |
Immunogen | Full length native protein purified from human muscle. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
Protein Aliases: CMD1IFLJ41013 CSM1 CSM2 desmin FLJ12025 FLJ39719 FLJ41793 intermediate filament protein; Desmin; intermediate filament protein; mutant desmin p.K241E
Gene Aliases: CSM1; CSM2; DES; LGMD2R
UniProt ID: (Human) P17661
Entrez Gene ID: (Human) 1674
Molecular Function: cytoskeletal protein intermediate filament structural protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.