Invitrogen
Manufacturer Code:PA517182
Catalog # PIPA517182
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to residues near the carboxy terminus of human desmin |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Desmin; intermediate filament protein; mutant desmin p.K241E
Gene Aliases: CSM1; CSM2; DES; LGMD2R
UniProt ID: (Human) P17661, (Rat) P48675, (Mouse) P31001
Entrez Gene ID: (Human) 1674, (Rat) 64362, (Mouse) 13346
Molecular Function:
cytoskeletal protein
intermediate filament
structural protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.