Proteintech
Manufacturer Code:105371AP150UL
Catalog # 50-172-6386
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Dermatopontin Fusion Protein Ag0801 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Dermatopontin; Dermatopontin DPT TRAMP; Early quiescence protein 1; early quiescence protein-1; EQ-1; TRAMP; Tyrosine-rich acidic matrix protein
Gene Aliases: 1810032B19Rik; 5033416F05Rik; DPT; EQ-1; Eq1; TRAMP
UniProt ID: (Human) Q07507, (Mouse) Q9QZZ6
Entrez Gene ID: (Human) 1805, (Mouse) 56429, (Rat) 289178
Molecular Function:
extracellular matrix protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.