Novus Biologicals
Manufacturer Code:NBP2847800.1ML
Catalog # NB032708
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the C terminal region of human Derlin-2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: carcinoma related; carcinoma related Degradation in endoplasmic reticulum protein 2 Der1-like domain family member 2 Der1-like protein 2 DER2 derlin-2 F-LAN-1DERtrin-2 FLANa F-LANaCGI-101; Degradation in endoplasmic reticulum protein 2; Der1-like domain family, member 2; Der1-like protein 2; Derlin-2; DERtrin-2; F-LAN-1; F-LANa
Gene Aliases: CGI-101; DER2; DERL2; DERtrin-2; F-LAN-1; F-LANa; FLANA; SBBI53
UniProt ID: (Human) Q9GZP9
Entrez Gene ID: (Human) 51009
Molecular Function: receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.