Invitrogen
Manufacturer Code:14588481
Catalog # 50-132-70
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Functional Assay (FN) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 33D1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: C-type lectin domain family 4, member a4; DC Marker DC inhibitor receptor 2 DCIR2; dendritic cell inhibitory receptor 2
Gene Aliases: Dcir2
Entrez Gene ID: (Mouse) 474145
Molecular Function:
cell adhesion molecule
cytokine receptor
defense/immunity protein
immunoglobulin receptor superfamily
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.