Proteintech
Manufacturer Code:168081AP150UL
Catalog # 50-173-1190
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Dematin Fusion Protein Ag10300 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Dematin; Dematin actin-binding protein; Dematin DMT DMTN EPB49; Erythrocyte membrane protein band 4.9; erythrocyte membrane protein band 4.9 (dematin); erythrocyte protein band 4.9
Gene Aliases: AI325486; dematin; DMT; DMTN; Epb4.9; EPB49
UniProt ID: (Human) Q08495, (Mouse) Q9WV69
Entrez Gene ID: (Human) 2039, (Rat) 361069, (Mouse) 13829
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.