Invitrogen
Manufacturer Code:PA527370
Catalog # PIPA527370
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 1 and 192 of Human Decorin |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Bone proteoglycan II; CSCD DSPG2 PG40 PGII PGS2 SLRR1B PG-S2 bone proteoglycan II decorin proteoglycan dermatan sulphate proteoglycans II proteoglycan core protein small leucine-rich protein 1B; Decorin; dermatan sulphate proteoglycans II; PG-S2; PG40; proteoglycan core protein; small leucine-rich protein 1B
Gene Aliases: CSCD; DCN; DSPG2; PG40; PGII; PGS2; SLRR1B
UniProt ID: (Human) P07585
Entrez Gene ID: (Human) 1634
Molecular Function:
cytokine
extracellular matrix protein
receptor
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.