Invitrogen
Manufacturer Code:PA513538
Catalog # PIPA513538
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 122-150 amino acids from the central region of human DCN |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Bone proteoglycan II; Decorin; Decorin Bone proteoglycan II PG-S2 PG40 SLRR1B DCN; dermatan sulphate proteoglycans II; PG-S2; PG40; proteoglycan core protein; small leucine-rich protein 1B
Gene Aliases: CSCD; DC; DCN; DSPG2; mDcn; PG40; PGII; PGS2; SLRR1B
UniProt ID: (Human) P07585, (Mouse) P28654
Entrez Gene ID: (Human) 1634, (Mouse) 13179
Molecular Function:
cytokine
extracellular matrix protein
receptor
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.