R&D Systems
Manufacturer Code:AF143SP
Catalog # AF143SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | <EM>S. frugiperda</EM> insect ovarian cell line <EM>Sf</EM> 21-derived recombinant human Decorin Gly17-Lys359 Accession # P07585 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: Bone proteoglycan II; Bone proteoglycan II CSCD DCN decorin decorin proteoglycan dermatan sulphate proteoglycans II DSPG2 PG40 PGII PG-II PGS2 PG-S2 proteoglycan core protein SLRR1B SLRR1BPGS2 small leucine-rich protein 1B; Decorin; dermatan sulphate proteoglycans II; PG-S2; PG40; proteoglycan core protein; small leucine-rich protein 1B
Gene Aliases: CSCD; DCN; DSPG2; PG40; PGII; PGS2; SLRR1B
UniProt ID: (Human) P07585
Entrez Gene ID: (Human) 1634
Molecular Function:
cytokine
extracellular matrix protein
receptor
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.