Invitrogen
Manufacturer Code:PA519924
Catalog # PIPA519924
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A peptide corresponding to amino acids near the amino terminus of human DcR3 precursor. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DcR3; Decoy receptor 3; decoy receptor 3 variant 1; decoy receptor 3 variant 2; Decoy receptor for Fas ligand; hCG&lowbar2044836 DCR3 DJ583P15.1.1 M68 M68E TR6 decoy receptor 3 decoy receptor for Fas ligand tumor necrosis factor receptor superfamily member 6B; M68; Tumor necrosis factor receptor superfamily member 6B; tumor necrosis factor receptor superfamily member 6b, decoy; tumor necrosis factor receptor superfamily, member 6b, decoy
Gene Aliases: DCR3; DJ583P15.1.1; M68; M68E; TNFRSF6B; TR6; UNQ186/PRO212
UniProt ID: (Human) O95407
Entrez Gene ID: (Human) 8771
Molecular Function: cytokine receptor receptor tumor necrosis factor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.