Invitrogen
Manufacturer Code:PA511808
Catalog # PIPA511808
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 1068-1097 amino acids from the central region of CAEEL Dcr-1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Death-promoting deoxyribonuclease; Endoribonuclease dcr-1; Endoribonuclease dcr-1 dcr-1; tDCR-1
Gene Aliases: CELE_K12H4.8; dcr-1; eri-4; K12H4.8; let-740
UniProt ID: (C. elegans) P34529
Entrez Gene ID: (C. elegans) 176138
Molecular Function:
DNA binding protein
endodeoxyribonuclease
nuclease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.