Invitrogen
Manufacturer Code:MA514974
Catalog # PIMA514974
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | K.180.3 |
Immunogen | Synthetic peptide corresponding to a region surrounding Gln255 of Daxx |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: CENP-C binding protein; CENP-C binding protein ETS1-associated protein 1 Fas-binding protein death domain-associated protein 6 death-associated protein 6 fas death domain-associated protein hDaxx; Daxx; Death domain-associated protein 6; death-associated protein 6; death-domain associated protein; EAP1; ETS1-associated protein 1; Fas death domain-associated protein; Fas-binding protein; hDaxx
Gene Aliases: BING2; DAP6; DAXX; EAP1
UniProt ID: (Human) Q9UER7, (Mouse) O35613
Entrez Gene ID: (Dog) 474873, (Bovine) 504336, (Human) 1616, (Mouse) 13163, (Rat) 140926
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.