Novus Biologicals
Manufacturer Code:NBP177011
Catalog # NBP177011
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Antibody was raised against a peptide corresponding to amino acids near the carboxy terminus of human Daxx. The immunogen is located within the last 50 amino acids of Daxx. Amino Acid Squence: TSVATQCDPEEIIVLSDSD |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BING2 CENP-C binding protein DAP6MGC126245 Daxx death domain-associated protein 6 death-associated protein 6 death-domain associated protein EAP1 ETS1-associated protein 1 Fas death domain-associated protein Fas-binding protein hDaxx MGC126246; CENP-C binding protein; Daxx; Death domain-associated protein 6; death-associated protein 6; death-domain associated protein; EAP1; ETS1-associated protein 1; Fas death domain-associated protein; Fas-binding protein; hDaxx
Gene Aliases: BING2; DAP6; DAXX; EAP1
UniProt ID: (Human) Q9UER7
Entrez Gene ID: (Human) 1616
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.