Abnova
Manufacturer Code:PAB0169
Catalog # 89-113-996
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to amino acids 44-57 of mouse Dap3. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of Dap3.
For Research Use Only
Protein Aliases: 28S ribosomal protein S29, mitochondrial; 4921514D13Rik; DAP-3; Death-associated protein 3; MRP-S29
Gene Aliases: 4921514D13Rik; DAP-3; Dap3; MRP-S29; Mrps29; S29mt
UniProt ID: (Mouse) Q9ER88
Entrez Gene ID: (Mouse) 65111
Molecular Function: RNA binding protein nucleic acid binding ribosomal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.