Invitrogen
Manufacturer Code:PA520629
Catalog # PIPA520629
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | a 12 amino acid peptide from near the amino terminus of human DACT2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Dapper antagonist of catenin 2; Dapper homolog 2; dapper homolog 2, antagonist of beta-catenin; dapper, antagonist of beta-catenin, homolog 2; dishevelled-binding antagonist of beta-catenin 2; mDpr2; PP13671 C6orf116 DAPPER2 DPR2 RP11-503C24.7 bA503C24.7 dapper homolog 2 dapper homolog 2 antagonist of beta-catenin
Gene Aliases: 2900084M21Rik; A630024E20; bA503C24.7; C6orf116; DACT2; DAPPER2; DPR2; Frd2; PP13671
UniProt ID: (Human) Q5SW24, (Mouse) Q7TN08
Entrez Gene ID: (Human) 168002, (Rat) 308212, (Mouse) 240025
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.