Abnova
Manufacturer Code:PAB9857
Catalog # 89-134-459
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant GST fusion protein corresponding to amino acids 1-179 of mouse Dab1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Sheep polyclonal antibody raised against partial recombinant Dab1.
For Research Use Only
Protein Aliases: AI956902; C630028C02Rik; Disabled homolog 1; scm; scr; scrambler; yot; yotari
Gene Aliases: AI956902; C630028C02Rik; Dab1; scm; scr; scrambler; yot
UniProt ID: (Mouse) P97318
Entrez Gene ID: (Mouse) 13131
Molecular Function:
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.