Invitrogen
Manufacturer Code:PA518517
Catalog # PIPA518517
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (PLQVSDYSWQQTKT) corresponding to the N-terminus amino acids of DYX1C1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: Dynein assembly factor 4, axonemal; dynein, axonemal, assembly factor 4; Dyslexia susceptibility 1 candidate gene 1 protein; DYX1C1 EKN1 dyslexia susceptibility 1 candidate 1 DYX1 DYXC1 FLJ37882 MGC70618
Gene Aliases: CILD25; DNAAF4; DYX1; DYX1C1; DYXC1; EKN1; RD
UniProt ID: (Human) Q8WXU2
Entrez Gene ID: (Human) 161582
Molecular Function:
chaperone
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.