Invitrogen
Manufacturer Code:PA5106353
Catalog # PIPA5106353
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human DYX1C1 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1700010I24Rik b2b811Clo CILD25 DNAAF4 Dynein assembly factor 4 axonemal dynein axonemal assembly factor 4 dyslexia susceptibility 1 candidate 1 dyslexia susceptibility 1 candidate 1 homolog dyslexia susceptibility 1 candidate 1 homolog (human) dyslexia susceptibility 1 candidate gene 1 protein Dyslexia susceptibility 1 candidate gene 1 protein homolog DYX1 Dyx1c1 DYXC1 Edem2 Ekn1 FLJ37882 MGC70618 RD; Dynein assembly factor 4, axonemal; dynein, axonemal, assembly factor 4; dyslexia susceptibility 1 candidate 1 homolog; Dyslexia susceptibility 1 candidate gene 1 protein; Dyslexia susceptibility 1 candidate gene 1 protein homolog
Gene Aliases: 1700010I24Rik; b2b811Clo; CILD25; DNAAF4; DYX1; DYX1C1; DYXC1; Edem2; EKN1; RD
UniProt ID: (Human) Q8WXU2, (Rat) Q5VJS5, (Mouse) Q8R368
Entrez Gene ID: (Human) 161582, (Rat) 363096, (Mouse) 67685
Molecular Function:
chaperone
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.