Novus Biologicals
Manufacturer Code:NBP247476
Catalog # NBP247476
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against a recombinant protein corresponding to amino acids: KLKEDSIPAPRSVGSIKINFTPRVFPTALRESQVAEEEEWLHKQAEARRAMNTDIAELCDLKEEEKNPEWLKDKGNKLFATENYLAAINA |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Dynein assembly factor 4, axonemal; dynein, axonemal, assembly factor 4; dyslexia susceptibility 1 candidate 1 dyslexia susceptibility 1 candidate gene 1 protein DYX1 DYXC1 EKN1MGC70618 FLJ37882 RD; Dyslexia susceptibility 1 candidate gene 1 protein
Gene Aliases: CILD25; DNAAF4; DYX1; DYX1C1; DYXC1; EKN1; RD
UniProt ID: (Human) Q8WXU2
Entrez Gene ID: (Human) 161582
Molecular Function: chaperone
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.