Abnova
Manufacturer Code:PAB6746
Catalog # 89-132-653
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to human DYX1C1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Goat polyclonal antibody raised against synthetic peptide of DYX1C1.
For Research Use Only
Protein Aliases: Dynein assembly factor 4, axonemal; dynein, axonemal, assembly factor 4; Dyslexia susceptibility 1 candidate gene 1 protein; DYX1; DYXC1; EKN1; FLJ37882; MGC70618; RD
Gene Aliases: CILD25; DNAAF4; DYX1; DYX1C1; DYXC1; EKN1; RD
UniProt ID: (Human) Q8WXU2
Entrez Gene ID: (Human) 161582
Molecular Function: chaperone
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.