Invitrogen
Manufacturer Code:PA514465
Catalog # PIPA514465
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 455-485 amino acids from the C-terminal region of human DYRK4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: dual specificity tyrosine-(Y)-phosphorylation regulated kinase 4; Dual specificity tyrosine-phosphorylation-regulated kinase 4; dual-specificity tyrosine-(Y)-phosphorylation regulated kinase 4; Dual-specificity tyrosine-phosphorylation regulated kinase 4
Gene Aliases: DYRK4
UniProt ID: (Human) A8K8F7
Entrez Gene ID: (Human) 8798
Molecular Function:
kinase
non-receptor serine/threonine protein kinase
non-receptor tyrosine protein kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.