Invitrogen
Manufacturer Code:PA5100911
Catalog # PIPA5100911
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human DYRK4 corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AW049118 dual specificity tyrosine phosphorylation regulated kinase 4 dual specificity tyrosine-(Y)-phosphorylation regulated kinase 4 dual specificity tyrosine-phosphorylation-regulated kinase 4 dual-specificity tyrosine-(Y)-phosphorylation regulated kinase 4 dyrk4 Dyrk4a Dyrk4b RAD51 associated protein 1 Rad51ap1; dual specificity tyrosine-(Y)-phosphorylation regulated kinase 4; Dual specificity tyrosine-phosphorylation-regulated kinase 4; dual-specificity tyrosine-(Y)-phosphorylation regulated kinase 4; dyrk4
Gene Aliases: AW049118; DYRK4; Dyrk4a; Dyrk4b
UniProt ID: (Human) A8K8F7, (Mouse) Q8BI55
Entrez Gene ID: (Human) 8798, (Mouse) 101320
Molecular Function:
kinase
non-receptor serine/threonine protein kinase
non-receptor tyrosine protein kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.