Invitrogen
Manufacturer Code:PA5111727
Catalog # PIPA5111727
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Protein corresonding to Human DYRK3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BC006704 Dual specificity tyrosine-phosphorylation-regulated kinase 3 Dyrk3 DYRK5 hYAK3-2 KSR RED REDK Regulatory erythroid kinase RP11-343H5.2 RSU2; dual specificity tyrosine-(Y)-phosphorylation regulated kinase 3; Dual specificity tyrosine-phosphorylation-regulated kinase 3; dual-specificity tyrosine-(Y)-phosphorylation regulated kinase 3; dual-specificity tyrosine-(Y)-phosphorylation regulated kinase 5; protein kinase Dyrk3; REDK; Regulatory erythroid kinase
Gene Aliases: DYRK3; DYRK5; hYAK3-2; RED; REDK
UniProt ID: (Human) O43781
Entrez Gene ID: (Human) 8444
Molecular Function: kinase non-receptor serine/threonine protein kinase non-receptor tyrosine protein kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.