R&D Systems
Manufacturer Code:AF5409
Catalog # AF5409
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | <EM>E. coli</EM>-derived recombinant human DYRK3 Asn52-Glu167 Accession # O43781 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: dual specificity tyrosine-(Y)-phosphorylation regulated kinase 3; Dual specificity tyrosine-phosphorylation-regulated kinase 3; dual specificity tyrosine-phosphorylation-regulated kinase 3 dual-specificity tyrosine-(Y)-phosphorylation regulated kinase 3 dual-specificity tyrosine-(Y)-phosphorylation regulated kinase 5 DYRK5 EC 2.7.12 EC 2.7.12.1 hYAK3-2 protein kinase Dyrk3 REDK REDKRED Regulatory erythroid kinase YAK3-2; dual-specificity tyrosine-(Y)-phosphorylation regulated kinase 3; dual-specificity tyrosine-(Y)-phosphorylation regulated kinase 5; protein kinase Dyrk3; REDK; Regulatory erythroid kinase
Gene Aliases: DYRK3; DYRK5; hYAK3-2; RED; REDK
UniProt ID: (Human) O43781
Entrez Gene ID: (Human) 8444
Molecular Function: kinase non-receptor serine/threonine protein kinase non-receptor tyrosine protein kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.