R&D Systems
Manufacturer Code:AF5407
Catalog # AF5407
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | <EM>E. coli</EM>-derived recombinant human DYRK1A Arg438-Thr511 Accession # Q13627 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: dual specificity tyrosine-(Y)-phosphorylation regulated kinase 1A; Dual specificity tyrosine-phosphorylation-regulated kinase 1A; Dual specificity YAK1-related kinase; dual-specificity tyrosine-(Y)-phosphorylation regulated kinase 1A DYRK1 MNBEC 2.7.110EC 2.7.12 MNBH; hMNB; HP86; mnb protein kinase homolog hp86; MNB/DYRK protein kinase; MNBH; Protein kinase minibrain homolog; serine/threonine kinase MNB; serine/threonine-specific protein kinase
Gene Aliases: DYRK; DYRK1; DYRK1A; HP86; MNB; MNBH; MRD7
UniProt ID: (Human) Q13627
Entrez Gene ID: (Human) 1859
Molecular Function:
kinase
non-receptor serine/threonine protein kinase
non-receptor tyrosine protein kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.