Proteintech
Manufacturer Code:119541AP150UL
Catalog # 50-172-7590
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | DYNLT1 Fusion Protein Ag2557 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Activator of G-protein signaling 2; AGS2; CW 1 DYNLT1 Protein CW 1 TCTEL1 tctex 1 TCTEX1; Dynein light chain Tctex-type 1; dynein light chain Tctex-type 1D; dynein, light chain, Tctex-type 1; Protein CW-1; t-complex testis expressed 1; T-complex testis-specific protein 1; T-complex testis-specific protein 1 homolog; t-complex-associated-testis-expressed 1-like 1; TCTEX-1
Gene Aliases: AGS2; CW-1; DYNLT1; TCTEL1; TCTEX-1; TCTEX1
UniProt ID: (Human) P63172, (Rat) Q9Z336, (Mouse) P51807
Entrez Gene ID: (Human) 6993, (Rat) 83462, (Mouse) 21648
Molecular Function:
cytoskeletal protein
microtubule family cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.