Invitrogen
Manufacturer Code:PA570692
Catalog # PIPA570692
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the N-terminal region of human DYNLRB1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BITH DNCL2A DNLC2A ROBLD1; Bithoraxoid-like protein; BLP; Dynein light chain 2A, cytoplasmic; Dynein light chain roadblock-type 1; dynein, cytoplasmic, light chain 2A; dynein, cytoplasmic, light polypeptide 2A; dynein, light chain, roadblock-type 1; Dynein-associated protein Km23; dynein-associated protein RKM23; km23; Roadblock domain-containing protein 1; ROBL/LC7-like 1; robl/LC7-like protein
Gene Aliases: 2010012N15Rik; 2010320M17Rik; 9430076K19Rik; AV124457; BITH; BLP; DNCL2A; DNLC2A; DYNLRB1; HSPC162; km23; km23-1; ROBLD1; zgc:56335
UniProt ID: (Bovine) Q3T140, (Human) Q9NP97, (Rat) P62628, (Mouse) P62627
Entrez Gene ID: (Horse) 100069352, (Rabbit) 100346957, (Bovine) 513342, (Human) 83658, (Dog) 477200, (Guinea pig) 100725912, (Rat) 170714, (Mouse) 67068, (Zebrafish) 394163
Molecular Function: cytoskeletal protein microtubule family cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.