OriGene
Manufacturer Code:TA501507
Catalog # 50-167-2648
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1B9 |
Immunogen | Full length human recombit protein of human DYNC1LI1 produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Cytoplasmic dynein 1 light intermediate chain 1; DLC-A; DLC-A DNCLI1 LIC1; Dynein light chain A; Dynein light intermediate chain 1, cytosolic; dynein, cytoplasmic, light intermediate polypeptide 1; LIC1
Gene Aliases: DLC-A; DNCLI1; DYNC1LI1; LIC1
UniProt ID: (Human) Q9Y6G9
Entrez Gene ID: (Human) 51143
Molecular Function: cytoskeletal protein enzyme modulator microtubule family cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.