Proteintech
Manufacturer Code:123451AP150UL
Catalog # 50-172-7911
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | DYNC1H1 Fusion Protein Ag2999 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cytoplasmic dynein 1 heavy chain 1; Cytoplasmic dynein heavy chain 1; cytoplasmic dynein heavy chain polypeptide 1; DHC1 DHC1a DNCH1 Dnchc1 DNCL DNECL DYHC DYNC1H1 Dynein heavy chain cytosolic HL 3 KIAA0325 p22; Dynein heavy chain, cytosolic; dynein heavy chain, retrograde transport; dynein, cytoplasmic 1, heavy chain 1; dynein, cytoplasmic, heavy chain 1; dynein, cytoplasmic, heavy polypeptide 1; legs at odd angle; MAP 1C; sprawling
Gene Aliases: 9930018I23Rik; AI894280; CMT2O; DHC1; DHC1a; DNCH1; Dnchc1; DNCL; Dnec1; DNECL; DYHC; DYNC1H1; fk70a07; HL-3; KIAA0325; Loa; MAP1C; mKIAA0325; P22; SMALED1; Swl; wu:fk70a07
UniProt ID: (Human) Q14204, (Rat) P38650, (Mouse) Q9JHU4
Entrez Gene ID: (Human) 1778, (Rat) 29489, (Mouse) 13424, (Zebrafish) 335110
Molecular Function:
cytoskeletal protein
hydrolase
microtubule binding motor protein
microtubule family cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.