Invitrogen
Manufacturer Code:PA527965
Catalog # PIPA527965
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 223 and 550 of Human Dishevelled 2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: dishevelled 2 (homologous to Drosophila dsh); dishevelled, dsh homolog 2; Dishevelled-2; DSH homolog 2; DSH homolog 2 dishevelled 2 (homologous to Drosophila dsh) dishevelled-2 segment polarity protein dishevelled homolog DVL-2; Segment polarity protein dishevelled homolog DVL-2
Gene Aliases: DVL2
UniProt ID: (Human) O14641, (Mouse) Q60838
Entrez Gene ID: (Human) 1856, (Mouse) 13543
Molecular Function:
enzyme modulator
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.