Invitrogen
Manufacturer Code:PA523409
Catalog # PIPA523409
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide (84 AEGAHSDAGSQGTDSHTD 101) of DVL-1 protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: dishevelled 1; dishevelled 1 (homologous to Drosophila dsh); dishevelled, dsh homolog 1; Dishevelled-1; DSH homolog 1; RP5-890O3.5 DVL DVL1L1 DSH homolog 1 dishevelled 1 (homologous to Drosophila dsh) dishevelled-1 segment polarity protein dishevelled homolog DVL-1; Segment polarity protein dishevelled homolog DVL-1
Gene Aliases: DRS2; DVL; dvl-1; DVL1; DVL1L1; DVL1P1; mKIAA4029
UniProt ID: (Human) O14640, (Rat) Q9WVB9, (Mouse) P51141
Entrez Gene ID: (Human) 1855, (Rat) 83721, (Mouse) 13542
Molecular Function: enzyme modulator signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.