Invitrogen
Manufacturer Code:PA5102836
Catalog # PIPA5102836
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human DVL1 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: dishevelled 1; dishevelled 1 (homologous to Drosophila dsh); dishevelled 1 dishevelled 1 (homologous to Drosophila dsh) dishevelled segment polarity protein 1 dishevelled dsh homolog 1 dishevelled dsh homolog 1 (Drosophila) dishevelled-1 Dishevelled-1-like DRS2 DSH homolog 1 DSH homolog 1-like Dvl Dvl1 dvl-1 DVL1L1 DVL1P1 MGC54245 mKIAA4029 RP5-890O3.5 Segment polarity protein dishevelled homolog DVL-1; dishevelled, dsh homolog 1; Dishevelled-1; DSH homolog 1; Segment polarity protein dishevelled homolog DVL-1
Gene Aliases: DRS2; DVL; dvl-1; DVL1; DVL1L1; DVL1P1; mKIAA4029
UniProt ID: (Human) O14640, (Rat) Q9WVB9, (Mouse) P51141
Entrez Gene ID: (Human) 1855, (Rat) 83721, (Mouse) 13542
Molecular Function:
enzyme modulator
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.