Proteintech
Manufacturer Code:273841AP150UL
Catalog # 50-173-5656
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | DVL1 Fusion Protein Ag26055 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: dishevelled 1 (homologous to Drosophila dsh); Dishevelled 1 DSH homolog 1 DVL DVL1; dishevelled, dsh homolog 1; Dishevelled-1; DSH homolog 1; Segment polarity protein dishevelled homolog DVL-1
Gene Aliases: DRS2; DVL; DVL1; DVL1L1; DVL1P1; mKIAA4029
UniProt ID: (Human) O14640, (Mouse) P51141
Entrez Gene ID: (Human) 1855, (Mouse) 13542
Molecular Function: enzyme modulator signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.