Invitrogen
Manufacturer Code:MA516147
Catalog # PIMA516147
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | P4H2 |
Immunogen | Synthetic peptide corresponding to the C-terminal of human DUX4. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: double homeobox 4-like; Double homeobox protein 10; Double homeobox protein 4; double homeobox protein 4/10; DUX10 double homeobox protein 10 double homeobox protein 4 double homeobox protein 4/10 double homeobox protein DUX10
Gene Aliases: DUX10; DUX4; DUX4L
UniProt ID: (Human) Q9UBX2
Entrez Gene ID: (Human) 100288687
Molecular Function:
DNA binding protein
helix-turn-helix transcription factor
homeobox transcription factor
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.