R&D Systems
Manufacturer Code:MAB9535100
Catalog # MAB9535100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 2142A |
Immunogen | Human DUX4 synthetic peptide |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: double homeobox 4 double homeobox protein 10 double homeobox protein 4 double homeobox protein 4/10 double homeobox protein DUX10 DUX10 DUX4L1; double homeobox 4-like; Double homeobox protein 10; Double homeobox protein 4; double homeobox protein 4/10
Gene Aliases: DUX10; DUX4; DUX4L
UniProt ID: (Human) Q9UBX2
Entrez Gene ID: (Human) 100288687
Molecular Function:
DNA binding protein
helix-turn-helix transcription factor
homeobox transcription factor
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.