Invitrogen
Manufacturer Code:PA579175
Catalog # PIPA579175
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence at the C-terminus of human DUT. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Deoxyuridine 5&prime-triphosphate nucleotidohydrolase mitochondrial dUTPase dUTP pyrophosphatase DUT; Deoxyuridine 5'-triphosphate nucleotidohydrolase; Deoxyuridine 5'-triphosphate nucleotidohydrolase, mitochondrial; dUTP diphosphatase; dUTP nucleotidohydrolase; dUTP pyrophosphatase; dUTPase; PIP4; PPAR-interacting protein 4
Gene Aliases: 5031412I06Rik; 5133400F09Rik; D2Bwg0749e; DUT; Dutp; dUTPase; PIP4
UniProt ID: (Human) P33316, (Rat) P70583
Entrez Gene ID: (Human) 1854, (Rat) 497778, (Mouse) 110074
Molecular Function: hydrolase phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.