Novus Biologicals
Manufacturer Code:NBP270610G
Catalog # NBP270610G
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI7C11 |
Immunogen | Recombinant protein fragment corresponding to amino acids 1-257 of human DUSP4 (NP_476499) produced in E.coli. |
Conjugate | DyLight 488 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: dual specificity phosphatase 4 Dual specificity protein phosphatase hVH2 EC 3.1.3.16 EC 3.1.3.48 HVH2serine/threonine specific protein phosphatase MAP kinase phosphatase 2 Mitogen-activated protein kinase phosphatase 2 MKP-2dual specificity protein phosphatase 4 MKP2VH1 homologous phosphatase 2 TYP VH2; Dual specificity protein phosphatase 4; Dual specificity protein phosphatase hVH2; MAP kinase phosphatase 2; Mitogen-activated protein kinase phosphatase 2; serine/threonine specific protein phosphatase; VH1 homologous phosphatase 2
Gene Aliases: DUSP4; HVH2; MKP-2; MKP2; TYP; VH2
UniProt ID: (Human) Q13115
Entrez Gene ID: (Human) 1846
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.