Abnova
Manufacturer Code:PAB4129
Catalog # 89-131-285
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide (conjugated with KLH) corresponding to C-terminus of human DUSP4. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of DUSP4.
For Research Use Only
Protein Aliases: Dual specificity protein phosphatase 4; Dual specificity protein phosphatase hVH2; HVH2; MAP kinase phosphatase 2; Mitogen-activated protein kinase phosphatase 2; MKP-2; MKP2; serine/threonine specific protein phosphatase; TYP; VH1 homologous phosphatase 2
Gene Aliases: DUSP4; HVH2; MKP-2; MKP2; TYP; VH2
UniProt ID: (Human) Q13115
Entrez Gene ID: (Human) 1846
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.