Abnova
Manufacturer Code:H00001845M07
Catalog # 89-021-370
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 2C4 |
Immunogen | DUSP3 (NP_004081 76 a.a. ~ 185 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a partial recombinant DUSP3.
For Research Use Only
Protein Aliases: Dual specificity protein phosphatase 3; Dual specificity protein phosphatase VHR; serine/threonine specific protein phosphatase; Vaccinia H1-related phosphatase; vaccinia virus phosphatase VH1-related; VHR
Gene Aliases: DUSP3; VHR
UniProt ID: (Human) P51452
Entrez Gene ID: (Human) 1845
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.