Invitrogen
Manufacturer Code:PA579174
Catalog # PIPA579174
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence at the C-terminus of human DUSP3. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: dual specificity phosphatase 3 (vaccinia virus phosphatase VH1-related)-like; Dual specificity protein phosphatase 3; Dual specificity protein phosphatase 3 Dual specificity protein phosphatase VHR Vaccinia H1-related phosphatase VHR DUSP3 VHR; Dual specificity protein phosphatase VHR; serine/threonine specific protein phosphatase; T-DSP11; Vaccinia H1-related phosphatase; vaccinia virus phosphatase VH1-related; VHR
Gene Aliases: 2210015O03Rik; 5031436O03Rik; DUSP3; RGD1560049; T-DSP11; VHR
UniProt ID: (Human) P51452, (Mouse) Q9D7X3
Entrez Gene ID: (Human) 1845, (Mouse) 72349, (Rat) 498003
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.