Invitrogen
Manufacturer Code:PA522001
Catalog # PIPA522001
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 1 and 185 of DUSP3 (Uniprot IDNo.P51452) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Dual specificity protein phosphatase 3; Dual specificity protein phosphatase VHR; serine/threonine specific protein phosphatase; T-DSP11; Vaccinia H1-related phosphatase; vaccinia virus phosphatase VH1-related; VHR; VHR dual specificity protein phosphatase 3 dual specificity protein phosphatase VHR serine/threonine specific protein phosphatase vaccinia H1-related phosphatase vaccinia virus phosphatase VH1-related
Gene Aliases: 2210015O03Rik; 5031436O03Rik; DUSP3; T-DSP11; VHR
UniProt ID: (Human) P51452, (Mouse) Q9D7X3
Entrez Gene ID: (Human) 1845, (Mouse) 72349
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.