Invitrogen
Manufacturer Code:PA522013
Catalog # PIPA522013
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to a region within amino acids 22 and 117 of DUSP26 (Uniprot IDNo.Q9BV47) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DSP-4; Dual specificity phosphatase SKRP3; Dual specificity protein phosphatase 26; dual-specificity phosphatase SKRP3; DUSP24 LDP-4 MKP8 NATA1 SKRP3 DSP-4 MAP kinase phosphatase 8 MKP-8 Novel amplified gene in thyroid anaplastic cancer dual specificity phosphatase SKRP3 dual specificity protein phosphatase 26 dual-specificity phosphatase SKRP3 low-molecular-mass dual-specificity phosphatase 4 mitogen-activated protein kinase phosphatase 8; Low-molecular-mass dual-specificity phosphatase 4; MAP kinase phosphatase 8; Mitogen-activated protein kinase phosphatase 8; neuroendocrine-associated phosphatase; Novel amplified gene in thyroid anaplastic cancer
Gene Aliases: 2310043K02Rik; DSP-4; DUSP24; DUSP26; LDP-4; LDP4; MKP-8; MKP8; NATA1; NEAP; SKRP3
UniProt ID: (Human) Q9BV47, (Mouse) Q9D700
Entrez Gene ID: (Human) 78986, (Mouse) 66959
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.