Proteintech
Manufacturer Code:165141AP150UL
Catalog # 50-173-1000
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | DUSP22 Fusion Protein Ag9734 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Dual specificity protein phosphatase 22; DUSP22 JKAP JNK stimulatory phosphatase 1 JSP 1 JSP1 LMW DSP2 LMWDSP2 MAP kinase phosphatase x MKP x MKPX VHX; homolog of mouse dual specificity phosphatase LMW-DSP2; JNK-stimulating phosphatase 1; JNK-stimulatory phosphatase-1; JSP-1; LMW-DSP2; Low molecular weight dual specificity phosphatase 2; MAP kinase phosphatase x; Mitogen-activated protein kinase phosphatase x
Gene Aliases: DUSP22; JKAP; JSP-1; JSP1; LMW-DSP2; LMWDSP2; MKP-x; MKPX; VHX
UniProt ID: (Human) Q9NRW4, (Mouse) Q99N11
Entrez Gene ID: (Human) 56940, (Mouse) 105352
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.