Proteintech
Manufacturer Code:129241AP150UL
Catalog # 50-172-8349
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | DUSP19 Fusion Protein Ag3611 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Dual specificity phosphatase TS-DSP1; Dual specificity protein phosphatase 19; DUSP17 DUSP19 LMW DSP3 LMWDSP3 Protein phosphatase SKRP1 SKRP1 TS DSP1; LMW-DSP3; Low molecular weight dual specificity phosphatase 3; Protein phosphatase SKRP1; SAPK pathway-regulating phosphatase 1; Stress-activated protein kinase pathway-regulating phosphatase 1
Gene Aliases: 5930436K22Rik; C79103; DUSP17; DUSP19; LMWDSP3; SKRP1; TS-DSP1
UniProt ID: (Human) Q8WTR2
Entrez Gene ID: (Human) 142679, (Mouse) 68082, (Rat) 311151
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.