Novus Biologicals
Manufacturer Code:NBP287312
Catalog # NB034934
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the C-terminal region of human DUSP11 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: dual specificity phosphatase 11 (RNA/RNP complex 1-interacting); dual specificity phosphatase 11 (RNA/RNP complex 1-interacting) Dual specificity protein phosphatase 11 EC 3.1.3.- Phosphatase that interacts with RNA/RNP complex 1 PIR1RNA/RNP complex-interacting phosphatase RNA/RNP complex-1-interacting phosphatase serine/threonine specific protein phosphatase; Dual specificity protein phosphatase 11; Phosphatase that interacts with RNA/RNP complex 1; RNA/RNP complex 1-interacting; RNA/RNP complex-1-interacting phosphatase; RNA/RNP complex-interacting phosphatase; serine/threonine specific protein phosphatase
Gene Aliases: DUSP11; PIR1
UniProt ID: (Human) O75319
Entrez Gene ID: (Human) 8446
Molecular Function:
hydrolase
nucleotidyltransferase
phosphatase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.